Learn More
eMolecules​ 1207760-37-9 | 4-Isoquinolinzinc bromide, 0.50 M in THF | Synthonix273.440 | C9H6BrNZn | 0.000 | [Br-].[Zn+]c1cncc2ccccc12 | 50ml | 532652879
Catalog No. 502696113 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Isoquinolinzinc bromide, 0.50 M in THF | Synthonix | 1207760-37-9273.440 | C9H6BrNZn | 0.000 | [Br-].[Zn+]c1cncc2ccccc12 | 50ml | 532652879
Catalog No. 50-269-6113 Supplier eMolecules​ Supplier No. 532652879
4-Isoquinolinzinc bromide, 0.50 M in THF | Synthonix | 1207760-37-9273.440 | C9H6BrNZn | 0.000 | [Br-].[Zn+]c1cncc2ccccc12 | 50ml | 532652879