Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isothiocyanato-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 97.0+%, TCI America™
SDP

Catalog No. I0486100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0486100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0486100MG Supplier TCI America Supplier No. I0486100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 36410-81-8
Melting Point 128°C
Molecular Formula C10H17N2OS
Quantity 100 mg
MDL Number MFCD00010109
Synonym 4-isothiocyanato-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-isothiocyanato-tempo, 4-isothiocyanato-2,2,6,6-tetramethylpiperidinooxyl, 4-itc-tempo, 4-isothiocyanato-1, 4-isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, 4-isothiocyanato-2,2,6,6-tetramethylpiperidinooxy radical, 1-piperidinyloxy, 4-isothiocyanato-2,2,6,6-tetramethyl, 4-isothiocyanato-tempo, for esr-spectroscopy, 4-isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical
InChI Key BKENZAFIBRCMKD-UHFFFAOYSA-N
SMILES CC1(C)CC(CC(C)(C)N1[O])N=C=S
IUPAC Name (4-isothiocyanato-2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Molecular Weight (g/mol) 213.32
PubChem CID 147645
Formula Weight 213.32
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Isothiocyanato-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical
Show More Show Less
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.