Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isothiocyanatobenzonitrile, 97%, Thermo Scientific™

Catalog No. AC348970050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC348970050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC348970050 Supplier Thermo Scientific Chemicals Supplier No. 348970050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2719-32-6
Molecular Formula C8H4N2S
Molecular Weight (g/mol) 160.194
InChI Key DZFKAXLNKZXNHD-UHFFFAOYSA-N
Synonym 4-cyanophenyl isothiocyanate, benzonitrile, 4-isothiocyanato, p-cyanophenyl isothiocyanate, 4-cyanophenylisothiocyanate, 4-cyano-phenyl-isothiocyanate, 4-isothiocyanatobenzenenitrile, 4-isothiocyanato-benzonitrile, benzonitrile,4-isothiocyanato, isothiocyanic acid, p-cyanophenyl ester, benzenenitrile, 4-isothiocyanato
PubChem CID 17626
IUPAC Name 4-isothiocyanatobenzonitrile
SMILES C1=CC(=CC=C1C#N)N=C=S

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 160.2
Chemical Name or Material 4-Isothiocyanatobenzonitrile
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.