Learn More
AdipoGen 4-ITCB-MPO-pyridinium bromide
SDP
Catalog No. 502560670
Encompass_Preferred
Chemical. CAS 155862-90-1. Formula C23H18BrN3O2S. MW 480.38. Synthetic. New class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance. Major disadvantage rel. low extinction coefficient.
Catalog No. 50-256-0670 Supplier AdipoGen Supplier No. CDXI0015M250
Chemical. CAS 155862-90-1. Formula C23H18BrN3O2S. MW 480.38. Synthetic. New class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance. Major disadvantage rel. low extinction coefficient.