Learn More
AdipoGen 4-ITCPE-MPO-pyridinium tosylat
SDP
Catalog No. 502560655
Encompass_Preferred
Chemical. CAS 155862-93-4. Formula C31H27N3O6S2. MW 601.69. Synthetic. Class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient Epsilon∼ >20,000 in EtOH.
Catalog No. 50-256-0655 Supplier AdipoGen Supplier No. CDXI0033M050
Chemical. CAS 155862-93-4. Formula C31H27N3O6S2. MW 601.69. Synthetic. Class of highly photostable, water soluble fluorescent labels with large Stokes shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient Epsilon∼ >20,000 in EtOH.