Learn More
Pro Scientific Inc 4 LITER BAG - LAB ARMOR BEADS
SDP
Catalog No. NC1777155
Encompass
Lab Armor Beads, by their design, provide a concurrent thermal and antimicrobial activity that efficiently shields the lab and personnel from invading organisms while thermally heating or cooling like water in any standard water bath, heat block or ice bucket.
Catalog No. NC1777155 Supplier Pro Scientific Inc Supplier No. B10011
Lab Armor Beads, by their design, provide a concurrent thermal and antimicrobial activity that efficiently shields the lab and personnel from invading organisms while thermally heating or cooling like water in any standard water bath, heat block or ice bucket.