Learn More
Alkali Scientific 4 Liter Lab Armor Metal Beads for Bead Bath
SDP
Catalog No. 502336206
Encompass_Preferred
4 Liter Lab Armor Metal Beads have a metal composition, are moisture and gas impermeable. They have high thermal conductivity, and a smooth, rounded surface. The working temperature range is -80°C to 180°C. They improve energy efficiency of standard water baths by over 50%.
Catalog No. 50-233-6206 Supplier Alkali Scientific Supplier No. WB525
4 Liter Lab Armor Metal Beads have a metal composition, are moisture and gas impermeable. They have high thermal conductivity, and a smooth, rounded surface. The working temperature range is -80°C to 180°C. They improve energy efficiency of standard water baths by over 50%.