Learn More
Invivogen QUANTI-Luc™ 4 Lucia/Gaussia, Highly Sensitive, Reliable Bioluminescence Detection, 500 ml
SDP
Catalog No. NC2470225
Encompass
QUANTI-Luc™ 4 Lucia/Gaussia is an advanced bioluminescence detection reagent that enables precise quantitative measurement of Lucia and Gaussia luciferase reporter gene expression. This ready-to-use solution ensures high sensitivity and reproducibility with exceptional signal-to-noise ratios, making it ideal for gene expression studies and reporter assays. Its unique formulation minimizes background interference and improves detection reliability, setting it apart in the field of molecular biology research.
Catalog No. NC2470225 Supplier Invivogen Supplier No. REPQLC4LG20
QUANTI-Luc™ 4 Lucia/Gaussia is an advanced bioluminescence detection reagent that enables precise quantitative measurement of Lucia and Gaussia luciferase reporter gene expression. This ready-to-use solution ensures high sensitivity and reproducibility with exceptional signal-to-noise ratios, making it ideal for gene expression studies and reporter assays. Its unique formulation minimizes background interference and improves detection reliability, setting it apart in the field of molecular biology research.