Learn More
Medchemexpress LLC 4-Maleimidobenzoic a 10g
SDP
Catalog No. 5000206832
Encompass_Preferred
4-Maleimidobenzoic acid is a protein crosslinker 4-Maleimidobenzoic acid can be used to synthesis maleimidobenzoyl spacers[1]
Catalog No. 50-002-06832 Supplier Medchemexpress LLC Supplier No. HYW10484810G
Add to Cart
Add to Cart
4-Maleimidobenzoic acid is a protein crosslinker 4-Maleimidobenzoic acid can be used to synthesis maleimidobenzoyl spacers[1]