Learn More
Cayman Chemical 4-MAPB (hydrochloride), Purity: ≥98%, 10 mg
SDP
Catalog No. 502894810
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; intended for research and forensic applications
Catalog No. 50-289-4810 Supplier Cayman Chemical Supplier No. 900210310 MG
An analytical reference standard categorized as a benzofuran; intended for research and forensic applications