Learn More
Bellco Glass, Inc. 4 Marine Prop With 1/2" Bore
SDP
Catalog No. 501944859
Encompass_Preferred
4" od 316l stainless steel, ra32 finish or betterMarine impeller with 1/2" bore & set screw. MADE IN THE USA.
Catalog No. 50-194-4859 Supplier Bellco Glass, Inc. Supplier No. 196473000
4" od 316l stainless steel, ra32 finish or betterMarine impeller with 1/2" bore & set screw. MADE IN THE USA.