Learn More
MOLECULAR DEPOT LLC 4-MEP BSA Conjugate - 1 mg
Catalog No. 5000013453
Encompass_Preferred
4-Mercapto-ethyl-pyridine (4-MEP) BSA Conjugate (1 mg) consists of 4-Mercapto-ethyl-pyridine (4-MEP) a chemical compound with a pyridine ring and a mercaptoethyl group conjugated to bovine serum albumin (BSA) through covalent bonding enhancing solubility and stability The 4-MEP BSA conjugate is studied for its interactions with biomolecules For in vitro research use only
Catalog No. 50-000-13453 Supplier MOLECULAR DEPOT LLC Supplier No. B2021262
4-Mercapto-ethyl-pyridine (4-MEP) BSA Conjugate (1 mg) consists of 4-Mercapto-ethyl-pyridine (4-MEP) a chemical compound with a pyridine ring and a mercaptoethyl group conjugated to bovine serum albumin (BSA) through covalent bonding enhancing solubility and stability The 4-MEP BSA conjugate is studied for its interactions with biomolecules For in vitro research use only