Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Mercaptopyridine

Catalog No. 501882815
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-188-2815 1 g
14-820-210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-188-2815 Supplier Sigma Aldrich Supplier No. 1482021G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Mercaptopyridine was used as a probe molecule to study the performance of the gold nanorod (AuNR)/polycaprolactone (PCL) fibrous mesh.
TRUSTED_SUSTAINABILITY

Specifications

CAS 4556-23-4
Molecular Formula C5H5NS
Linear Formula C5H5NS
Quantity 1 g
MDL Number MFCD00006422
Synonym 4-Pyridinethiol
Molecular Weight (g/mol) 111.16
Percent Purity 95%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.