Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methanesulfonamido)benzoic acid, Thermo Scientific™

Catalog No. AC469660050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469660050 5 g
AC469660010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC469660050 Supplier Thermo Scientific Chemicals Supplier No. 469660050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 4-(Methanesulfonamido)benzoic acid
Alpha Vector METHANESULFONAMIDOBENZOICACID
CAS 7151-76-0
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C8 H9 N O4 S
Quantity 5 g
MDL Number 00025052
Synonym 4-[(Methylsulfonyl)amino]benzoic acid
Molecular Weight (g/mol) 215.23
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.