Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methanesulfonamido)benzoic acid, Thermo Scientific™

Catalog No. p-7227413
Click to view available options
Quantity:
1 g
5 g

Specifications

Alpha Vector METHANESULFONAMIDOBENZOICACID
Chemical Name or Material 4-(Methanesulfonamido)benzoic acid
CAS 7151-76-0
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C8 H9 N O4 S
MDL Number 00025052
Quantity 1 g
Synonym 4-[(Methylsulfonyl)amino]benzoic acid
Molecular Weight (g/mol) 215.23
Physical Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.