Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-1-naphthaldehyde 99%, Thermo Scientific™

Catalog No. AC146940500
Change view
Click to view available options
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC146940500 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC146940500 Supplier Thermo Scientific Chemicals Supplier No. 146940500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15971-29-6
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C12H10O2
Linear Formula CH3OC10H6CHO
Quantity 50g
MDL Number MFCD00004006
Beilstein 08, 147
Synonym 4-methoxy-1-naphthaldehyde, 4-methoxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 4-methoxy, 4-methoxynaphthalenecarbaldehyde, pubchem16658, acmc-209dk5, ksc494i2d, 1-naphthaldehyde, 4-methoxy, 4-methoxy-1-naphthalenecarbaldehyde
InChI Key MVXMNHYVCLMLDD-UHFFFAOYSA-N
SMILES COC1=CC=C(C2=CC=CC=C21)C=O
IUPAC Name 4-methoxynaphthalene-1-carbaldehyde
Molecular Weight (g/mol) 186.21
PubChem CID 85217
Formula Weight 186.21
Percent Purity ≥98.5%
Chemical Name or Material 4-Methoxy-1-naphthaldehyde, 99%
Show More Show Less
EINECSNumber 240-109-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.