Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-1-naphthalenemethanol 96.0+%, TCI America™
SDP

Catalog No. M20861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20861G 1 g
M20865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20861G Supplier TCI America Supplier No. M20861G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16820-54-5
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD04089264
InChI Key VBXSDYNXURBEQH-UHFFFAOYSA-N
Synonym 1-(Hydroxymethyl)-4-methoxynaphthalene
PubChem CID 900419
IUPAC Name (4-methoxynaphthalen-1-yl)methanol
SMILES COC1=CC=C(C2=CC=CC=C21)CO

Specifications

Melting Point 76°C
Quantity 1 g
Formula Weight 188.23
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-1-naphthalenemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.