Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-2,6-dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M22445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2244-5G 5 g
M22441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2244-5G Supplier TCI America Supplier No. M22445G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 4-Methoxy-2,6-dimethylphenylboronic Acid
CAS 361543-99-9
Quantity 5 g
Packaging Glass Bottle
Physical Form Crystal-Powder at 20°C
Molecular Formula C9H13BO3
MDL Number MFCD03452755
Synonym 2,6-dimethyl-4-methoxyphenyl boronic acid, 2,6-dimethyl-4-methoxyphenylboronic acid, 4-methoxy-2,6-dimethylphenyl boronic acid, 2,6-dimethyl-4-methoxybenzeneboronic acid, 2-borono-5-methoxy-m-xylene
InChI Key UOSSBXMFWPYHEF-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1C)OC)C)(O)O
IUPAC Name (4-methoxy-2,6-dimethylphenyl)boronic acid
Molecular Weight (g/mol) 180.01
PubChem CID 3716303
Formula Weight 180.01
Percent Purity ≥98.0%
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.