Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxy-2-methylbenzaldehyde, 95%

Click to view available options
Quantity:
1g
5g
52289-54-0
C9H10O2
150.177
MFCD02261771
WICYVKGMEJSDAO-UHFFFAOYSA-N
2-methyl-4-methoxybenzaldehyde,4-methoxy-2-methyl-benzaldehyde,benzaldehyde, 4-methoxy-2-methyl,2-methyl-p-anisaldehyde,2-methyl-anisaldehyde,acmc-1aoyo,4-methoxy-2-methylbenzaldehyd,4-methoxy-2-methylbenzaldehyde,3-furane-2-amido benzoic acid,4-methoxy-2-methylbenzaldehyde
283285
4-methoxy-2-methylbenzaldehyde
CC1=C(C=CC(=C1)OC)C=O
52289-54-0
C9H10O2
150.177
MFCD02261771
WICYVKGMEJSDAO-UHFFFAOYSA-N
2-methyl-4-methoxybenzaldehyde,4-methoxy-2-methyl-benzaldehyde,benzaldehyde, 4-methoxy-2-methyl,2-methyl-p-anisaldehyde,2-methyl-anisaldehyde,acmc-1aoyo,4-methoxy-2-methylbenzaldehyd,4-methoxy-2-methylbenzaldehyde,3-furane-2-amido benzoic acid,4-methoxy-2-methylbenzaldehyde
283285
4-methoxy-2-methylbenzaldehyde
CC1=C(C=CC(=C1)OC)C=O
Quantity 5g
Refractive Index 1.5710
Density 1.107g/mL
Boiling Point 262°C to 267°C (literature)
Color Yellow
Beilstein 1933786
Formula Weight 150.18
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 4-Methoxy-2-methylbenzaldehyde