Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-2-methylphenyl isocyanate, 97%, Thermo Scientific™

Catalog No. AAL1208603
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1208603 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1208603 Supplier Thermo Scientific Chemicals Supplier No. L1208603

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 60385-06-0
Density 1.121
Boiling Point 74°C to 76°C (1mmHg)
Flash Point >110°C (230°F)
Molecular Formula C9H9NO2
Refractive Index 1.544
Quantity 1g
MDL Number MFCD00013852
UN Number UN2206
Beilstein 7115803
Sensitivity Moisture sensitive
Synonym 4-methoxy-2-methylphenyl isocyanate, 4-methoxy-2-methylphenylisocyanate, benzene, 1-isocyanato-4-methoxy-2-methyl, acmc-1bfcu, 4-methoxy-o-tolyl isocyanate, 4-methoxy-2-methylbenzenisocyanate, 1-isocyanato-4-methoxy-2-methyl benzene, 1-isocyanato-4-methoxy-2-methyl-benzene, benzene,1-isocyanato-4-methoxy-2-methyl
InChI Key XGRQPTXFVDRJMY-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)OC)N=C=O
IUPAC Name 1-isocyanato-4-methoxy-2-methylbenzene
Molecular Weight (g/mol) 163.176
PubChem CID 2734905
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 4-Methoxy-2-methylphenyl isocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 624-219-1
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.