Learn More
Medchemexpress LLC 4-methoxy-3-[[2-piperidin-1-yl-5-(trifluoromethyl)phenyl]sulfamoyl]benzoic acid | 865273-97-8 | MFCD07309764 | 98.8% | C20H21F3N2O5S | 10MG
SDP
Catalog No. 5000258566
Encompass_Preferred
ERAP1-IN-1 is a small-molecule research compound that selectively inhibits endoplasmic reticulum aminopeptidase 1 (ERAP1). It competitively inhibits ERAP1 activity against nonamer peptide substrates and is supplied at high purity for biochemical and cellular studies of antigen processing and peptide trimming.
  • Selective ERAP1 inhibition enabling studies of antigen processing
  • Competitive inhibition against nonamer peptide substrates
  • High reported purity (98.77%) suitable for research assays
  • Available in multiple small-scale pack sizes for laboratory experiments
  • Molecular weight 458.45 and defined molecular formula for experiment planning
Catalog No. 50-002-58566 Supplier Medchemexpress LLC Supplier No. HY13312510MG
ERAP1-IN-1 is a small-molecule research compound that selectively inhibits endoplasmic reticulum aminopeptidase 1 (ERAP1). It competitively inhibits ERAP1 activity against nonamer peptide substrates and is supplied at high purity for biochemical and cellular studies of antigen processing and peptide trimming.
  • Selective ERAP1 inhibition enabling studies of antigen processing
  • Competitive inhibition against nonamer peptide substrates
  • High reported purity (98.77%) suitable for research assays
  • Available in multiple small-scale pack sizes for laboratory experiments
  • Molecular weight 458.45 and defined molecular formula for experiment planning