Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-3,5-dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M26855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2685-5G 5 g
M26851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2685-5G Supplier TCI America Supplier No. M26855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 301699-39-8 | C9H13BO3 | 180.01 g/mol

Specifications

Chemical Name or Material 4-Methoxy-3,5-dimethylphenylboronic Acid
CAS 301699-39-8
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C9H13BO3
MDL Number MFCD01114648
Synonym 3,5-dimethyl-4-methoxyphenylboronic acid, 3,5-dimethyl-4-methoxybenzeneboronic acid, 4-methoxy-3,5-dimethylphenyl boronic acid, 4-methoxy-3,5-dimethyl-phenyl boronic acid, 4-methoxy-3,5-dimethylbenzeneboronic acid
InChI Key WZUCSPWZHRVOSD-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C(=C1)C)OC)C)(O)O
IUPAC Name (4-methoxy-3,5-dimethylphenyl)boronic acid
Molecular Weight (g/mol) 180.01
PubChem CID 3817586
Formula Weight 180.01
Percent Purity 97.0 to 112.0%
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.