Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-3,5-dinitrotoluene 98.0+%, TCI America™
SDP

Catalog No. M18425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18425G Supplier TCI America Supplier No. M18425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29455-11-6
Molecular Formula C8H8N2O5
Molecular Weight (g/mol) 212.16
MDL Number MFCD00192097
InChI Key HVNUPXQONRHUOX-UHFFFAOYSA-N
Synonym 4-Methyl-2,6-dinitroanisole, 1-Methoxy-4-methyl-2,6-dinitrobenzene
PubChem CID 259649
IUPAC Name 2-methoxy-5-methyl-1,3-dinitrobenzene
SMILES COC1=C(C=C(C)C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 124°C
Quantity 5 g
UN Number 2930
Formula Weight 212.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-3,5-dinitrotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.