Learn More
Sigma Aldrich 4-Methoxy-3-(morpholinomethyl)benzaldehyde
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Methylglyoxal is a toxic endogenous by-product of glycolysis
- It is a reactive dicarbonyl compound that promotes non-enzymatic glycation of proteins to yield irreversible advanced glycated end products, leading to the cross-linking or degradation of proteins.methylglyoxal solution is used in cytotoxic studies
Specifications
Specifications
| CAS | 128501-81-5 |
| Molecular Formula | C13H17NO3 |
| Linear Formula | C13H17NO3 |
| MDL Number | MFCD00442737 |
| Quantity | 100 mL |
| Molecular Weight (g/mol) | 235.28 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.