Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-4-Methoxy-Alpha-Methylbenzylamine

Catalog No. 72684225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-684-225G 25 g
72-684-2100G 100 g
72-684-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 72-684-225G Supplier Sigma Aldrich Supplier No. 72684225G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(+)-4-Methoxy-+a-methylbenzylamine can be used as a reactant to prepare: Enantiopure stereoisomers of hemicryptophanes, which are used for the recognition of glucopyranosides. Bicyclic Geissman-Waiss lactone via intramolecular ring-closure reaction of the diastereomeric mixture of sulfonium salts. N-[(1R)-1-(4-Methoxyphenyl)ethyl]-N'sa-methylthiourea by reacting with methyl isothiocyanate.ChIpr∗

Specifications

CAS 22038-86-4
Density 1.024 g/mL at 20°C
Molecular Formula C9H13NO
Linear Formula C9H13NO
MDL Number MFCD00671659
Quantity 25 g
Synonym (R)-(+)-1-(4-Methoxyphenyl)ethylamine
Molecular Weight (g/mol) 151.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.