Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methoxy-N-[2-[(E)-2-pyridin-4-ylethenyl]phenyl]benzenesulfonamide | 173528-92-2 | MFCD00945104 | 97.1% | 366.43 g/mol | C20H18N2O3S | 25 MG
SDP

Catalog No. 5000247202
Encompass_Preferred

HMN-154 is a benzenesulfonamide research compound (CAS 173528-92-2) supplied as a high-purity solid for in vitro anticancer studies. It shows potent activity against KB and colon38 cell lines and is provided in pre-measured quantities for laboratory use.

  • High purity solid suitable for cellular assays.
  • Reported potent cytotoxic activity against KB and colon38 cells (IC50 0.0026-0.003 μg/mL).
  • Available in small, pre-weighed quantities for research use.
  • Molecular formula C20H18N2O3S; molecular weight 366.43 g/mol.
  • CAS registry number 173528-92-2 for unambiguous identification.

Catalog No. 50-002-47202 Supplier Medchemexpress LLC Supplier No. HY10300125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HMN-154 is a benzenesulfonamide research compound (CAS 173528-92-2) supplied as a high-purity solid for in vitro anticancer studies. It shows potent activity against KB and colon38 cell lines and is provided in pre-measured quantities for laboratory use.

  • High purity solid suitable for cellular assays.
  • Reported potent cytotoxic activity against KB and colon38 cells (IC50 0.0026-0.003 μg/mL).
  • Available in small, pre-weighed quantities for research use.
  • Molecular formula C20H18N2O3S; molecular weight 366.43 g/mol.
  • CAS registry number 173528-92-2 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.