Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methoxy-n-[[4-(trifluoromethyl)phenyl]methyl]butanamide | 202402-01-5 | MFCD31812537 | 98.7% | 275.27 | C13H16F3NO2 | 25 MG
SDP

Catalog No. 5000242975
Encompass_Preferred

GET73 is a research chemical (a γ-hydroxybutyric acid analog) studied for anxiolytic and anti-alcohol effects and for modulation of glutamate transmission in the hippocampus. Intended for laboratory research use only; not for human or veterinary use.

  • High purity 98.7% suitable for research applications.
  • Molecular weight 275.27 for accurate dosing calculations.
  • Chemical formula C13H16F3NO2 for structure-based studies.
  • Available as solid and as 10 mM solution in DMSO.
  • Used in studies of alcohol-related behavior and glutamate modulation.

Catalog No. 50-002-42975 Supplier Medchemexpress LLC Supplier No. HY10803425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GET73 is a research chemical (a γ-hydroxybutyric acid analog) studied for anxiolytic and anti-alcohol effects and for modulation of glutamate transmission in the hippocampus. Intended for laboratory research use only; not for human or veterinary use.

  • High purity 98.7% suitable for research applications.
  • Molecular weight 275.27 for accurate dosing calculations.
  • Chemical formula C13H16F3NO2 for structure-based studies.
  • Available as solid and as 10 mM solution in DMSO.
  • Used in studies of alcohol-related behavior and glutamate modulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.