Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-trans-stilbene 98.0+%, TCI America™
SDP

Catalog No. M29641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29641G 1 g
M29645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29641G Supplier TCI America Supplier No. M29641G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1694-19-5
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00017178
InChI Key XWYXLYCDZKRCAD-BQYQJAHWSA-N
PubChem CID 1245920
ChEBI CHEBI:35094
IUPAC Name 1-methoxy-4-[(E)-2-phenylethenyl]benzene
SMILES COC1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Melting Point 136°C
Color White
Boiling Point 126°C
Quantity 1 g
Formula Weight 210.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-trans-stilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.