Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Methoxyacetanilide, 99+%, Thermo Scientific™

Catalog No. AC249711000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC249711000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC249711000 Supplier Thermo Scientific Chemicals Supplier No. 249711000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-66-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
InChI Key XVAIDCNLVLTVFM-UHFFFAOYSA-N
Synonym n-4-methoxyphenyl acetamide, 4'-methoxyacetanilide, p-methoxyacetanilide, p-acetanisidide, p-acetanisidine, methacetin, 4-methoxyacetanilide, aceto-p-anisidide, n-acetyl-p-anisidine, acetamide, n-4-methoxyphenyl
PubChem CID 5827
IUPAC Name N-(4-methoxyphenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)OC

Specifications

Melting Point 129.0°C to 132.0°C
Linear Formula CH3C6H4NHC(O)CH3
Quantity 100 g
Solubility Information Solubility in water: 0.42g/100mL (20°C). Other solubilities: soluble in diluted acids and alkalies, soluble in alcohol, acetone and chloroform, 7g/100mL water (100°C), insoluble in non-polar solvents
Formula Weight 165.19
Chemical Name or Material 4'-Methoxyacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.