Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Methoxybenzenesulfonyl fluoride

Catalog No. ALD0013010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD0013010G 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ALD0013010G Supplier Sigma Aldrich Supplier No. ALD0013010G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-chloro-2-methylbutane is a halogenated organic building block
  • It can be synthesized by employing 2-methylbutanol as starting reagent
  • Grignard reagent derived from (+)-1-chloro-2-methylbutane may be employed for the preparation of partially optically active (-)-alkylphenylcarbinols and partially optically active secondary carbinols
  • Infrared and raman spectra of its optically active form (+)(s)-1-chloro-2-methylbutane in various phases (liquid, glass and crystal) have been evaluated

Specifications

CAS 368-91-2
Density 1.339 g/L (at 25°C)
Molecular Formula C7H7FO3S
Refractive Index n/D 1.154
Linear Formula C7H7FO3S
MDL Number MFCD21881374
Quantity 50 mL
Molecular Weight (g/mol) 190.19
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.