Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Methoxybenzyl isocyanate

Catalog No. 4874065G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-740-65G 5 g
48-740-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48-740-65G Supplier Sigma Aldrich Supplier No. 4874065G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methoxybenzyl isocyanate (pmbnco), also known as p-methoxybenzyl isocyanate, is an organic building block containing an isocyanate group. its enthalpy of vaporization at boiling point has been reported.4-methoxybenzyl isocyanate (p-methoxybenzyl isocyanate) may be used in the synthesis of 1-(4-methoxybenzyl)-3-{2-[(4-nitrophenyl)amino]ethyl}urea and 1-(4-methoxybenzyl)-3-{2-[(5-nitropyridin-2-yl)amino]ethyl} urea.it may be used in the multi-step synthesis of:
  • Diimidazodiazepine containing 5:7:5-fused heterocycle
  • 2-deoxy-2-ureido-d-ribo-hexulofuranosonamide
  • Bicyclic guanidine core of batzelladine a

Specifications

CAS 56651-60-6
Density 1.143 g/mL (at 25°C (literature))
Boiling Point 60°C to 66°C (0.6 mmHg)
Molecular Formula C9H9NO2
Refractive Index n20/D 1.433 (literature)
Linear Formula CH3OC6H4CH2NCO
MDL Number MFCD00673064
Quantity 5 g
Molecular Weight (g/mol) 163.17
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.