Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M190725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1907-25G 25 g
M19071G 1 g
M19075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1907-25G Supplier TCI America Supplier No. M190725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99768-12-4
Molecular Formula C8H9BO4
MDL Number MFCD01632203
InChI Key PQCXFUXRTRESBD-UHFFFAOYSA-N
PubChem CID 2734369

Specifications

Melting Point 234°C
Color White-Yellow
Quantity 25 g
Synonym methyl 4-boronobenzoate, 4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl phenyl boronic acid, 4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonylphenyl boronic acid
SMILES COC(=O)C1=CC=C(C=C1)B(O)O
IUPAC Name [4-(methoxycarbonyl)phenyl]boronic acid
Molecular Weight (g/mol) 179.97
Formula Weight 179.97
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.