Learn More
Medchemexpress LLC 4'-Methoxychalcone | 959-23-9 | 99.8% | 10 G
SDP
Catalog No. 5000342614
Encompass_Preferred
4'-Methoxychalcone regulates adipocyte differentiation through PPARγ activation. It also modulates the expression and secretion of various adipokines in adipose tissue that are involved in insulin sensitivity. This product is for research use only.
  • Regulates adipocyte differentiation through PPARγ activation.
  • Modulates the expression and secretion of various adipokines.
  • Increases mRNA expression of adipogenic genes during differentiation.
  • Reduces upregulated mRNA expression of inflammatory cytokines.
  • Suitable for research use.
Catalog No. 50-003-42614 Supplier Medchemexpress LLC Supplier No. HY12840010G
4'-Methoxychalcone regulates adipocyte differentiation through PPARγ activation. It also modulates the expression and secretion of various adipokines in adipose tissue that are involved in insulin sensitivity. This product is for research use only.
  • Regulates adipocyte differentiation through PPARγ activation.
  • Modulates the expression and secretion of various adipokines.
  • Increases mRNA expression of adipogenic genes during differentiation.
  • Reduces upregulated mRNA expression of inflammatory cytokines.
  • Suitable for research use.