Learn More
Medchemexpress LLC 4'-Methoxychalcone | 959-23-9 | 99.8% | 5 G
SDP
Catalog No. 5000341196
Encompass_Preferred
4'-Methoxychalcone regulates adipocyte differentiation through PPARγ activation. It also modulates the expression and secretion of various adipokines in adipose tissue that are involved in insulin sensitivity. This product is intended for research use only.
  • Regulates adipocyte differentiation via PPARγ activation
  • Modulates adipokine expression and secretion involved in insulin sensitivity
  • Increases mRNA expression of adipogenic genes during differentiation
  • Reduces upregulated mRNA expression of IL-6, PAI-1, and MCP-1
  • Causes an increase in PPARγ expression during differentiation
  • Exhibits antiproliferative activity and cytotoxicity against various human cell lines
  • Targets PPARγ
  • Derived from Piperaceae Piper nigrum Linn.
  • Classified as flavonoids, chalcones
Catalog No. 50-003-41196 Supplier Medchemexpress LLC Supplier No. HY1284005G
4'-Methoxychalcone regulates adipocyte differentiation through PPARγ activation. It also modulates the expression and secretion of various adipokines in adipose tissue that are involved in insulin sensitivity. This product is intended for research use only.
  • Regulates adipocyte differentiation via PPARγ activation
  • Modulates adipokine expression and secretion involved in insulin sensitivity
  • Increases mRNA expression of adipogenic genes during differentiation
  • Reduces upregulated mRNA expression of IL-6, PAI-1, and MCP-1
  • Causes an increase in PPARγ expression during differentiation
  • Exhibits antiproliferative activity and cytotoxicity against various human cell lines
  • Targets PPARγ
  • Derived from Piperaceae Piper nigrum Linn.
  • Classified as flavonoids, chalcones