Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxychalcone 98.0+%, TCI America™
SDP

Catalog No. M140925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M140925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M140925G Supplier TCI America Supplier No. M140925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 959-33-1
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00017179
InChI Key XUFXKBJMCRJATM-FMIVXFBMSA-N
Synonym 4-methoxychalcone, e-3-4-methoxyphenyl-1-phenylprop-2-en-1-one, 2e-3-4-methoxyphenyl-1-phenylprop-2-en-1-one, chalcone, 4-methoxy, 3-4-methoxyphenyl-1-phenyl-2-propen-1-one, 2-propen-1-one, 3-4-methoxyphenyl-1-phenyl, 2-propen-1-one, 3-4-methoxyphenyl-1-phenyl-, 2e, phenyl p-methoxystyryl ketone, 4'-methoxybenzylideneacetophenone, 3-4-methoxyphenyl-1-phenylprop-2-en-1-one
PubChem CID 641819
IUPAC Name (E)-3-(4-methoxyphenyl)-1-phenylprop-2-en-1-one
SMILES COC1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 75°C
Color Yellow
Quantity 25 g
Formula Weight 238.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxychalcone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.