Learn More
Cayman Chemical 4-Methoxycinnamic Acid, Purity: ≥98%, CAS: 830-09-1, 50 g
SDP
Catalog No. 502916945
Encompass_Preferred
A phenolic acid with diverse biological activities; reduces A. fumigatus biofilm formation from 0.4-3.2 mM; reduces the number of dysplastic aberrant crypt foci and dysplastic liver cells in a rat model of 1,2-dimethylhydrazine-induced colon carcinogenesis at 80 mg/kg; reverses MK-801-induced decreases in the preference ratio for the novel object in the novel object recognition test in mice; reduces plasma glucose concentrations in a rat model of STZ-induced diabetes
Catalog No. 50-291-6945 Supplier Cayman Chemical Supplier No. 3950050 G
A phenolic acid with diverse biological activities; reduces A. fumigatus biofilm formation from 0.4-3.2 mM; reduces the number of dysplastic aberrant crypt foci and dysplastic liver cells in a rat model of 1,2-dimethylhydrazine-induced colon carcinogenesis at 80 mg/kg; reverses MK-801-induced decreases in the preference ratio for the novel object in the novel object recognition test in mice; reduces plasma glucose concentrations in a rat model of STZ-induced diabetes