Learn More
Medchemexpress LLC 4-Methoxycinnamaldehyde | 1963-36-6 | 99.89% | 162.19 | 50 MG
SDP
Catalog No. 5000454459
Encompass_Preferred
4-Methoxycinnamaldehyde (p-Methoxycinnamaldehyde) is an active constituent derived from Agastache rugosa. It demonstrates cytoprotective activity against the respiratory syncytial virus (RSV) in a human larynx carcinoma cell line. The compound effectively inhibits the cytopathic effect of RSV with an estimated IC50 of 0.055 μg/mL.
  • Demonstrates cytoprotective activity against the respiratory syncytial virus (RSV).
  • Effectively inhibits the cytopathic effect of RSV.
  • Derived from Agastache rugosa.
Catalog No. 50-004-54459 Supplier Medchemexpress LLC Supplier No. HYW013605A50MG
4-Methoxycinnamaldehyde (p-Methoxycinnamaldehyde) is an active constituent derived from Agastache rugosa. It demonstrates cytoprotective activity against the respiratory syncytial virus (RSV) in a human larynx carcinoma cell line. The compound effectively inhibits the cytopathic effect of RSV with an estimated IC50 of 0.055 μg/mL.
  • Demonstrates cytoprotective activity against the respiratory syncytial virus (RSV).
  • Effectively inhibits the cytopathic effect of RSV.
  • Derived from Agastache rugosa.