Learn More
Medchemexpress LLC 4-Methoxycinnamaldehyde | 1963-36-6 | 99.9% | 162.1852 | 500 MG
SDP
Catalog No. 5000454497
Encompass_Preferred
4-Methoxycinnamaldehyde (p-Methoxycinnamaldehyde), an active constituent of Agastache rugosa, exhibits cytoprotective activity against respiratory syncytial virus (RSV) in a human larynx carcinoma cell line. It effectively inhibits the cytopathic effect of RSV with an estimated IC50 of 0.055 μg/mL.
  • Exhibits cytoprotective activity against respiratory syncytial virus (RSV).
  • Effectively inhibits the cytopathic effect of RSV.
  • Light yellow to yellow solid appearance.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Insoluble in H2O.
  • Can be prepared in various in vivo dissolution methods.
  • For research use only.
Catalog No. 50-004-54497 Supplier Medchemexpress LLC Supplier No. HYW013605A500MG
4-Methoxycinnamaldehyde (p-Methoxycinnamaldehyde), an active constituent of Agastache rugosa, exhibits cytoprotective activity against respiratory syncytial virus (RSV) in a human larynx carcinoma cell line. It effectively inhibits the cytopathic effect of RSV with an estimated IC50 of 0.055 μg/mL.
  • Exhibits cytoprotective activity against respiratory syncytial virus (RSV).
  • Effectively inhibits the cytopathic effect of RSV.
  • Light yellow to yellow solid appearance.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Insoluble in H2O.
  • Can be prepared in various in vivo dissolution methods.
  • For research use only.