Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Methoxycinnamaldehyde | 1963-36-6 | 99.9% | 162.19 | 10 MG
SDP

Catalog No. 5000454207
Encompass_Preferred

4-Methoxycinnamaldehyde is an active constituent of *Agastache rugosa* that shows cytoprotective activity against respiratory syncytial virus (RSV) in human larynx carcinoma cell lines. It effectively inhibits the cytopathic effect of RSV with an estimated IC50 of 0.055 μg/mL.

  • For research use only
  • Appearance: solid
  • Color: light yellow to yellow
  • Initial source: plants (Araceae, Acorus gramineus Soland.)
  • Shipping: room temperature in continental US
  • Storage: 4°C, stored under nitrogen

Catalog No. 50-004-54207 Supplier Medchemexpress LLC Supplier No. HYW013605A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methoxycinnamaldehyde is an active constituent of *Agastache rugosa* that shows cytoprotective activity against respiratory syncytial virus (RSV) in human larynx carcinoma cell lines. It effectively inhibits the cytopathic effect of RSV with an estimated IC50 of 0.055 μg/mL.

  • For research use only
  • Appearance: solid
  • Color: light yellow to yellow
  • Initial source: plants (Araceae, Acorus gramineus Soland.)
  • Shipping: room temperature in continental US
  • Storage: 4°C, stored under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.