Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Methoxycinnamaldehyde | 1963-36-6 | 99.9% | 162.19 | 5 MG
SDP

Catalog No. 5000454477
Encompass_Preferred

4-Methoxycinnamaldehyde, also known as p-Methoxycinnamaldehyde, is an active component found in *Agastache rugosa*. It demonstrates cytoprotective effects against the respiratory syncytial virus (RSV) within human larynx carcinoma cell lines. The compound is effective in inhibiting the cytopathic effect of RSV, with an estimated IC50 of 0.055 μg/mL.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Structure classification: Ketones, Aldehydes, Acids
  • Initial source: Plants (Araceae Acorus gramineus Soland.)
  • Storage: 4°C, stored under nitrogen; in solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)

Catalog No. 50-004-54477 Supplier Medchemexpress LLC Supplier No. HYW013605A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methoxycinnamaldehyde, also known as p-Methoxycinnamaldehyde, is an active component found in *Agastache rugosa*. It demonstrates cytoprotective effects against the respiratory syncytial virus (RSV) within human larynx carcinoma cell lines. The compound is effective in inhibiting the cytopathic effect of RSV, with an estimated IC50 of 0.055 μg/mL.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Structure classification: Ketones, Aldehydes, Acids
  • Initial source: Plants (Araceae Acorus gramineus Soland.)
  • Storage: 4°C, stored under nitrogen; in solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.