Learn More
Medchemexpress LLC 4-Methoxycinnamaldehyde | 1963-36-6 | MFCD00017343 | 99.9% | 162.19 | 100 MG
SDP
Catalog No. 5000454655
Encompass_Preferred
4-Methoxycinnamaldehyde, an active constituent of *Agastache rugosa*, demonstrates cytoprotective activity against respiratory syncytial virus (RSV) in a human larynx carcinoma cell line. It effectively inhibits the cytopathic effect of RSV with an estimated IC50 of 0.055 μg/mL.
  • Active constituent of *Agastache rugosa*.
  • Exhibits cytoprotective activity against respiratory syncytial virus (RSV).
  • Effectively inhibits cytopathic effect of RSV.
Catalog No. 50-004-54655 Supplier Medchemexpress LLC Supplier No. HYW013605A100MG
4-Methoxycinnamaldehyde, an active constituent of *Agastache rugosa*, demonstrates cytoprotective activity against respiratory syncytial virus (RSV) in a human larynx carcinoma cell line. It effectively inhibits the cytopathic effect of RSV with an estimated IC50 of 0.055 μg/mL.
  • Active constituent of *Agastache rugosa*.
  • Exhibits cytoprotective activity against respiratory syncytial virus (RSV).
  • Effectively inhibits cytopathic effect of RSV.