Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Methoxycoumarin | 20280-81-3 | 176.17 g/mol | C10H8O3 | 1 G
SDP

Catalog No. 5000210755
Encompass_Preferred

4-Methoxycoumarin is a coumarin derivative used in research with reported antibacterial activity against Ralstonia solanacearum. It is supplied as a powder with chemical formula C10H8O3 (molecular weight 176.17 g/mol) and CAS number 20280-81-3. The compound is soluble in DMSO (100 mg/mL; ultrasonic assistance recommended), and storage guidance and solution stability are provided for laboratory handling.

  • Antibacterial activity against Ralstonia solanacearum
  • Chemical formula C10H8O3 and molecular weight 176.17 g/mol
  • CAS number 20280-81-3 for unambiguous identification
  • High DMSO solubility: 100 mg/mL (requires ultrasonic)
  • Storage stability: powder -20°C (up to 3 years), 4°C (up to 2 years); in solvent -80°C (6 months), -20°C (1 month)
  • Solution preparation guidance from 1 g: 1 mM = 5.6763 mL, 5 mM = 1.1353 mL, 10 mM = 0.5676 mL

Catalog No. 50-002-10755 Supplier Medchemexpress LLC Supplier No. HYW0676731G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methoxycoumarin is a coumarin derivative used in research with reported antibacterial activity against Ralstonia solanacearum. It is supplied as a powder with chemical formula C10H8O3 (molecular weight 176.17 g/mol) and CAS number 20280-81-3. The compound is soluble in DMSO (100 mg/mL; ultrasonic assistance recommended), and storage guidance and solution stability are provided for laboratory handling.

  • Antibacterial activity against Ralstonia solanacearum
  • Chemical formula C10H8O3 and molecular weight 176.17 g/mol
  • CAS number 20280-81-3 for unambiguous identification
  • High DMSO solubility: 100 mg/mL (requires ultrasonic)
  • Storage stability: powder -20°C (up to 3 years), 4°C (up to 2 years); in solvent -80°C (6 months), -20°C (1 month)
  • Solution preparation guidance from 1 g: 1 mM = 5.6763 mL, 5 mM = 1.1353 mL, 10 mM = 0.5676 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.