Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methoxycoumarin | 20280-81-3 | >98.0% | 176.17 g/mol | C10H8O3 | 100 MG
SDP

Catalog No. 5000210759
Encompass_Preferred

4-methoxycoumarin is a coumarin-class small molecule supplied for research use and reported to have antibacterial activity against Ralstonia solanacearum. It is described in supplier documentation with molecular formula C10H8O3, molecular weight 176.17 g/mol, and solubility in DMSO of 10 mM.

  • Antibacterial activity reported vs. Ralstonia solanacearum
  • Purity >98.0%
  • Molecular weight 176.17 g/mol
  • Chemical formula C10H8O3
  • Solubility 10 mM in DMSO
  • Intended for research use only

Catalog No. 50-002-10759 Supplier Medchemexpress LLC Supplier No. HYW067673100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-methoxycoumarin is a coumarin-class small molecule supplied for research use and reported to have antibacterial activity against Ralstonia solanacearum. It is described in supplier documentation with molecular formula C10H8O3, molecular weight 176.17 g/mol, and solubility in DMSO of 10 mM.

  • Antibacterial activity reported vs. Ralstonia solanacearum
  • Purity >98.0%
  • Molecular weight 176.17 g/mol
  • Chemical formula C10H8O3
  • Solubility 10 mM in DMSO
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.