Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methoxycoumarin | 20280-81-3 | MFCD00017349 | >99.0% | 176.17 g/mol | C10H8O3 | 25 MG
SDP

Catalog No. 5000209549
Encompass_Preferred

4-methoxycoumarin is a coumarin derivative reported to show antibacterial activity against R. solanacearum and is supplied for research use in small quantities for analytical and biological testing.

  • Shows reported antibacterial activity against R. solanacearum.
  • Molecular formula: C10H8O3.
  • Molecular weight: 176.17 g/mol.
  • CAS number: 20280-81-3.
  • Typical pack size: 25 mg; intended for research use.
  • Reported purity (supplier): >99%.

Catalog No. 50-002-09549 Supplier Medchemexpress LLC Supplier No. 25MGHYW067673
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-methoxycoumarin is a coumarin derivative reported to show antibacterial activity against R. solanacearum and is supplied for research use in small quantities for analytical and biological testing.

  • Shows reported antibacterial activity against R. solanacearum.
  • Molecular formula: C10H8O3.
  • Molecular weight: 176.17 g/mol.
  • CAS number: 20280-81-3.
  • Typical pack size: 25 mg; intended for research use.
  • Reported purity (supplier): >99%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.