Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxydiphenylamine 98.0+%, TCI America™
SDP

Catalog No. M20695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20695G 5 g
M206925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20695G Supplier TCI America Supplier No. M20695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1208-86-2
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.25
MDL Number MFCD00228649
InChI Key OBHGSIGHEBGGFS-UHFFFAOYSA-N
PubChem CID 14581
IUPAC Name 4-methoxy-N-phenylaniline
SMILES COC1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 106°C
Color White-Yellow
Boiling Point 195°C
Quantity 5 g
Formula Weight 199.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxydiphenylamine
EINECSNumber (3)-3450
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.