Learn More
Medchemexpress LLC 4'-Methoxyflavanone | 97005-76-0 | 99.5% | 254.285 g/mol | 5 MG
SDP
Catalog No. 5000426901
Encompass_Preferred
4'-Methoxyflavanone is a laboratory chemical for research use only. It is intended to be handled by suitably qualified and experienced scientists in appropriately equipped facilities. The product is provided as a solid with a melting point range of 155-158°C.
  • Identified uses include laboratory chemicals and manufacture of substances.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-004-26901 Supplier Medchemexpress LLC Supplier No. HYN92465MG
4'-Methoxyflavanone is a laboratory chemical for research use only. It is intended to be handled by suitably qualified and experienced scientists in appropriately equipped facilities. The product is provided as a solid with a melting point range of 155-158°C.
  • Identified uses include laboratory chemicals and manufacture of substances.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.