Learn More
Medchemexpress LLC 4'-Methoxyflavone | 4143-74-2 | 99.63% | 252.26 | 25 MG
SDP
Catalog No. 5000352489
Encompass_Preferred
4'-Methoxyflavone is a flavonoid with antioxidant properties and anti-proliferative activity against cancer cells. It can inhibit cycle-dependent kinases, leading to cell cycle arrest, and regulate cell signaling pathways to influence cell proliferation and apoptosis. This compound can be used in cancer chemoprevention research.
  • Flavonoid with antioxidant properties
  • Anti-proliferative activity against cancer cells
  • Inhibits cycle-dependent kinases
  • Leads to cell cycle arrest
  • Regulates cell signaling pathways to influence cell proliferation and apoptosis
  • Used in cancer chemoprevention research
Catalog No. 50-003-52489 Supplier Medchemexpress LLC Supplier No. 25MGHY139054
4'-Methoxyflavone is a flavonoid with antioxidant properties and anti-proliferative activity against cancer cells. It can inhibit cycle-dependent kinases, leading to cell cycle arrest, and regulate cell signaling pathways to influence cell proliferation and apoptosis. This compound can be used in cancer chemoprevention research.
  • Flavonoid with antioxidant properties
  • Anti-proliferative activity against cancer cells
  • Inhibits cycle-dependent kinases
  • Leads to cell cycle arrest
  • Regulates cell signaling pathways to influence cell proliferation and apoptosis
  • Used in cancer chemoprevention research