Learn More
eMolecules​ 27524-58-9 | [4-(Methoxymethoxy)phenyl]magnesium bromide, 0.50 M in THF | Synthonix241.367 | C8H9BrMgO2 | 0.000 | [Br-].COCOc1ccc([Mg+])cc1 | 100ml | 812262713
Catalog No. 502694452 Shop All eMolecules Inc. Products
Encompass_Preferred
[4-(Methoxymethoxy)phenyl]magnesium bromide, 0.50 M in THF | Synthonix | 27524-58-9241.367 | C8H9BrMgO2 | 0.000 | [Br-].COCOc1ccc([Mg+])cc1 | 100ml | 812262713
Catalog No. 50-269-4452 Supplier eMolecules​ Supplier No. 812262713
[4-(Methoxymethoxy)phenyl]magnesium bromide, 0.50 M in THF | Synthonix | 27524-58-9241.367 | C8H9BrMgO2 | 0.000 | [Br-].COCOc1ccc([Mg+])cc1 | 100ml | 812262713