Learn More
Medchemexpress LLC 4-Methoxyphenethyl alcohol | 702-23-8 | 152.19 | 1 G
SDP
Catalog No. 5000445871
Encompass_Preferred
4-Methoxyphenethyl alcohol is an aromatic alcohol and a major component in the anise-like odor produced by Amorphophallus. It inhibits the protein, RNA, and DNA synthesis in Escherichia coli.
  • Inhibits protein, RNA, and DNA synthesis in Escherichia coli.
  • Demonstrates complete bacteriostatic concentration of 4.8 mM against Escherichia coli.
  • Primary component of the anise-like odor produced by Amorphophallus.
  • Storage for pure form: -20°C for 3 years, 4°C for 2 years.
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-004-45871 Supplier Medchemexpress LLC Supplier No. HYW0040561G
4-Methoxyphenethyl alcohol is an aromatic alcohol and a major component in the anise-like odor produced by Amorphophallus. It inhibits the protein, RNA, and DNA synthesis in Escherichia coli.
  • Inhibits protein, RNA, and DNA synthesis in Escherichia coli.
  • Demonstrates complete bacteriostatic concentration of 4.8 mM against Escherichia coli.
  • Primary component of the anise-like odor produced by Amorphophallus.
  • Storage for pure form: -20°C for 3 years, 4°C for 2 years.
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month.