Learn More
Medchemexpress LLC 4-Methoxyphenethyl alcohol | 702-23-8 | 152.19 | 10 G
SDP
Catalog No. 5000449163
Encompass_Preferred
4-Methoxyphenethyl alcohol is an aromatic alcohol identified for laboratory chemical uses and manufacturing of substances. It is also known as Benzeneethanol, 4-methoxy-. It is a primary component of the anise-like odor produced by Amorphophallus and acts as an inhibitor of protein, RNA, and DNA synthesis in Escherichia coli. The appearance is colorless to off-white, solid below 26°C and liquid above 28°C.
  • High purity of 99.9%.
  • Colorless to off-white appearance, solid below 26°C and liquid above 28°C.
  • Molecular formula: C9H12O2.
  • Rigorous testing ensures compliance with specifications.
  • Identified for laboratory chemical uses and manufacturing of substances.
  • Inhibits protein, RNA, and DNA synthesis in Escherichia coli.
Catalog No. 50-004-49163 Supplier Medchemexpress LLC Supplier No. HYW00405610G
4-Methoxyphenethyl alcohol is an aromatic alcohol identified for laboratory chemical uses and manufacturing of substances. It is also known as Benzeneethanol, 4-methoxy-. It is a primary component of the anise-like odor produced by Amorphophallus and acts as an inhibitor of protein, RNA, and DNA synthesis in Escherichia coli. The appearance is colorless to off-white, solid below 26°C and liquid above 28°C.
  • High purity of 99.9%.
  • Colorless to off-white appearance, solid below 26°C and liquid above 28°C.
  • Molecular formula: C9H12O2.
  • Rigorous testing ensures compliance with specifications.
  • Identified for laboratory chemical uses and manufacturing of substances.
  • Inhibits protein, RNA, and DNA synthesis in Escherichia coli.